missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATAD2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25679-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a zinc finger protein transcriptional repressor. The encoded protein is part of the methyl-CpG-binding protein-1 complex, which represses gene expression by deacetylating methylated nucleosomes. Mutations in this gene are linked to intellectual disability and dysmorphic features associated with cognitive disability.Specifications
GATAD2B | |
Polyclonal | |
Unconjugated | |
GATAD2B | |
GATAD2B, KIAA1150, p66/p68, P66beta, RP11 216N14.6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57459 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WXI9 | |
GATAD2B | |
GATAD2B Fusion Protein Ag22615 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |