missing translation for 'onlineSavingsMsg'
Learn More

GATC Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen GATC
Concentration 0.17 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16882225
View Documents
Proteintech
23183-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16872225
View Documents
Proteintech
23183-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GATC (glutamyl-tRNA(Gln) amidotransferase, subunit C) is a protein-coding gene. GO annotations related to this gene include glutaminyl-tRNA synthase (glutamine-hydrolyzing) activity. An important paralog of this gene is ENSG00000111780.
Specifications

Specifications

GATC
Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
O43716, Q8CBY0
283459, 384281
GATC Fusion Protein Ag19713
Primary
-20°C
GATC
0.17 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
15E1.2, GATC, GatC like protein, Protein 15E1.2
GATC
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GATC Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.