Learn More
GBA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27972-1-AP-20UL
Description
The GBA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GBA. This antibody recognizes human, mouse antigen. The GBA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
GBA | |
Polyclonal | |
Unconjugated | |
20 μL | |
2629 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.