Learn More
GBA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27972-1-AP-150UL
 View more versions of this product
Product Code. 16875925
  • 4652.90 SEK / 150µL
Description
The GBA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GBA. This antibody recognizes human, mouse antigen. The GBA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GBA
Polyclonal
Unconjugated
150 μL
2629
IgG
Western Blot, Immunohistochemistry, ELISA
367 μg/mL
Rabbit
Primary
Human, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GBA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL