Läs mer
GBA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17164-1-AP-150UL
 Se fler versioner av denna produkt
Produktkod. 16886994
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.
Specifikationer
Specifikationer
GBA2
Polyclonal
Unconjugated
GBA2
AD035, Beta glucocerebrosidase 2, Beta glucosidase 2, GBA2, Glucosylceramidase 2, KIAA1605, NLGase
Rabbit
Antigen Affinity Chromatography
RUO
230101, 298399, 57704
-20°C
Liquid
ELISA
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5M868, Q69ZF3, Q9HCG7
GBA2
GBA2 Fusion Protein Ag10606
150 μL
Primary
Human, Mouse, Rat