missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | GBA2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896994
|
Proteintech
17164-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16886994
|
Proteintech
17164-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.Specifications
GBA2 | |
ELISA | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q5M868, Q69ZF3, Q9HCG7 | |
230101, 298399, 57704 | |
GBA2 Fusion Protein Ag10606 | |
Primary | |
-20°C | |
GBA2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AD035, Beta glucocerebrosidase 2, Beta glucosidase 2, GBA2, Glucosylceramidase 2, KIAA1605, NLGase | |
GBA2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |