missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17164-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.Specifications
GBA2 | |
Polyclonal | |
Unconjugated | |
GBA2 | |
AD035, Beta glucocerebrosidase 2, Beta glucosidase 2, GBA2, Glucosylceramidase 2, KIAA1605, NLGase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230101, 298399, 57704 | |
-20°C | |
Liquid |
ELISA | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5M868, Q69ZF3, Q9HCG7 | |
GBA2 | |
GBA2 Fusion Protein Ag10606 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |