Learn More
Thermo Scientific™ GC Syringe for Shimadzu
Brand: Thermo Scientific™ 365D6620
1118.27 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
23s G | |
10 μL | |
Shimadzu |
Ea. | |
Cone | |
Fixed |
Product Content Correction
Product Title
Thermo Scientific™ GC Syringe for Shimadzu > Syringe, 10 μL, For Shimadzu, Length: 42 mm, Gauge: 23S, Cone, 1/EA
Spot an opportunity for improvement?Share a Content Correction