Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand:  Trajan Scientific and Medical™ 062710
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12750740
  • 325.00 SEK / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
Tubing
0.025 mm
1 m
GC, LC and Biotechnology
0.363 mm