Learn More
GCLM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14241-1-AP-150UL
 View more versions of this product
Product Code. 16822794
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
Specifications
Specifications
GCLM
Polyclonal
Unconjugated
GCLM
Gamma ECS regulatory subunit, GCLM, GCS light chain, GLCLR
Rabbit
Antigen Affinity Chromatography
RUO
14630, 2730, 29739
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O09172, P48507, P48508
GCLM
GCLM Fusion Protein Ag5497
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GCLM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated