Learn More
GCM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21088-1-AP-150UL
 View more versions of this product
Product Code. 16840695
  • 4652.90 SEK / 150µL
Description
This gene is a homolog of the Drosophila glial cells missing gene, which is thought to act as a binary switch between neuronal and glial cell determination. The protein encoded by this gene contains a conserved N-terminal GCM motif that has DNA-binding activity. The protein is a transcription factor that acts as a master regulator of parathyroid development. It has been suggested that this transcription factor might mediate the effect of calcium on parathyroid hormone expression and secretion in parathyroid cells. Mutations in this gene are associated with hypoparathyroidism.
Specifications
Specifications
GCM2
Polyclonal
Unconjugated
GCM2
GCM motif protein 2, GCM2, GCMB, Glial cells missing homolog 2, hGCMb
Rabbit
Antigen Affinity Chromatography
RUO
107889, 9247
-20°C
Liquid
Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O09102, O75603
GCM2
GCM2 Fusion Protein Ag15334
150 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GCM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated