Learn More
GEFT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14839-1-AP-20UL
 View more versions of this product
Product Code. 16883694
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
Specifications
Specifications
GEFT
Polyclonal
Unconjugated
ARHGEF25
ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor
Rabbit
Antigen Affinity Chromatography
RUO
115557, 314904, 52666
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P720, Q86VW2, Q9CWR0
Arhgef25
Peptide
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GEFT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated