Learn More
Scientific Industries Genie Clip Plate
The Genie Clip Plate is an accessory for the Roto-Shake Genie, Enviro-Genie, Incubator Genie, and Rotator Genie.
Brand: Scientific Industries SI-1124
Additional Details : Weight : 0.10000kg
Description
- Stainless steel clip plate to secure tubes
- Choose to accommodate: 3 tubes (28 to 30 mm OD), 6 tubes (15 to 17 mm OD), or 12 microtubes
- Maximum of 8 clip plates, 4 per side, may be attached to the Magnetic Platform for use with the Roto-Shake Genie, Enviro-Genie or Incubator-Genie
- Maximum of 12 clip plates, 6 per side, may be attached to the Magnetic Platform for use with the Rotator Genie
Specifications
6 x 15 to 17 mm dia. Test tubes per Plate | |
Clip Plate | |
14 x 7.6 cm (5.5 x 3 in.) | |
6 |
15 to 17 mm dia. | |
Stainless Steel | |
Roto-Shake Genie, Enviro-Genie, Incubator Genie, and Rotator Genie | |
1 Ea. |