Läs mer
Invitrogen™ Gentamicin Monoclonal Antibody (GB73.3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46131
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA1-10725 detects Gentamicin. MA1-10725 has been successfully used in ELISA applications. The MA1-10725 immunogen is Gentamicin, conjugated to KLH.
Gentamicin is an aminoglycoside antibiotic, used to treat many types of bacterial infections, particularly those caused by Gram-negative bacteria. However, gentamicin is not used for Neisseria gonorrhoeae, Neisseria meningitidis or Legionella pneumophila bacterial infections (because of the risk of the patient going into shock from lipid A endotoxin found in certain gram negative organisms).
Specifikationer
| Gentamicin | |
| Monoclonal | |
| 1.3 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2a |
| ELISA | |
| GB73.3 | |
| Unconjugated | |
| Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7 | |
| Gentamicin, conjugated to KLH. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |