missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GFM2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896724
|
Proteintech
16941-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886724
|
Proteintech
16941-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFM2 is involved in mitochondrial translation, which is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintece of mitochondrial DNA. In Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. This gene encodes one of mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. Alternative splicing results in at least three transcript variants encoding distinct isoforms.Specifications
GFM2 | |
Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q5BJP6, Q8R2Q4, Q969S9 | |
294672, 320806, 84340 | |
GFM2 Fusion Protein Ag10530 | |
Primary | |
-20°C | |
GFM2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EF G2mt, EFG2, Elongation factor G2, GFM2, hEFG2, mEF G 2, MST027, RRF2mt | |
Gfm2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GFM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech