missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFRA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21973-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes all the three isoforms.
GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.Specifications
GFRA2 | |
Polyclonal | |
Unconjugated | |
GFRA2 | |
GDNF family receptor alpha 2, GDNF receptor alpha 2, GDNF receptor beta, GDNFR alpha 2, GDNFR beta, GDNFRB, GFR alpha 2, GFRA2, Neurturin receptor alpha, NRTNR ALPHA, NTNR alpha, NTNRA, RET ligand 2, RETL2, TRNR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14586, 2675 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00451, O08842 | |
Gfra2 | |
GFRA2 Fusion Protein Ag16893 | |
150 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |