missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFRA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25379-1-AP-20UL
This item is not returnable.
View return policy
Description
GFRa-3 binds persephin, thus, persephin, GFRa-3, and Ret PTK form a complex to transduce persephin signal and to mediate persephin function.Specifications
GFRA3 | |
Polyclonal | |
Unconjugated | |
GFRA3 | |
GDNF family receptor alpha 3, GDNF receptor alpha 3, GDNFR alpha 3, GFR alpha 3, GFRA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14587, 2676 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35118, O60609 | |
Gfra3 | |
GFRA3 Fusion Protein Ag13253 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GFRA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction