missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GIPC1 |
---|---|
Concentration | 0.41 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883664
|
Proteintech
14822-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873664
|
Proteintech
14822-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May be involved in G protein-linked signaling.Specifications
GIPC1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O14908, Q9Z0G0, Q9Z254 | |
10755, 67903, 83823 | |
GIPC1 Fusion Protein Ag6597 | |
Primary | |
-20°C | |
GIPC1 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C19orf3, GIPC, GIPC1, GLUT1CBP, Hs.6454, IIP 1, NIP, RGS GAIP interacting protein, RGS19 interacting protein 1, RGS19IP1, SEMCAP, SYNECTIIN, SYNECTIN, Tax interaction protein 2, TIP 2 | |
Gipc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GIPC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech