Learn More
GIYD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21158-1-AP-20UL
 View more versions of this product
Product Code. 16890765
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes a protein that is an important regulator of genome stability. The protein represents the catalytic subunit of the SLX1-SLX4 structure-specific endonuclease, which can resolve DNA secondary structures that are formed during repair and recombination processes. Two identical copies of this gene are located on the p arm of chromosome 16 due to a segmental duplication; this record represents the more telomeric copy. Alternative splicing results in multiple transcript variants. Read-through transcription also occurs between this gene and the downstream SULT1A4 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 4) gene.
Specifications
Specifications
GIYD2
Polyclonal
Unconjugated
SLX1B
GIY YIG domain containing 2, GIYD1, GIYD2, SLX1
Rabbit
Antigen Affinity Chromatography
RUO
75764, 79008
-20°C
Liquid
Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BX32, Q9BQ83
Slx1b
GIYD2 Fusion Protein Ag13094
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GIYD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated