missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11395-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The approximately 142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintece of telomere length.Specifications
GKAP1 | |
Polyclonal | |
Unconjugated | |
GKAP1 | |
FKSG21, G kinase anchoring protein 1, GKAP1, GKAP42 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361202, 56278, 80318 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VSY0, Q5XIG5, Q9JMB0 | |
Gkap1 | |
GKAP1 Fusion Protein Ag1914 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |