missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GKAP1 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828443
|
Proteintech
11395-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818443
|
Proteintech
11395-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The approximately 142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintece of telomere length.Spécification
GKAP1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q5VSY0, Q5XIG5, Q9JMB0 | |
361202, 56278, 80318 | |
GKAP1 Fusion Protein Ag1914 | |
Primary | |
-20°C | |
GKAP1 |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FKSG21, G kinase anchoring protein 1, GKAP1, GKAP42 | |
Gkap1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |