Learn More
ROBU Glass Adapter
Glass Adapter for use with a crucible.
Brand: ROBU 20802
Additional Details : Weight : 0.23280kg
Description
- Adapter
- For filter crucible
Specifications
For 50 mL Filter Crucibles | |
Borosilicate Glass (3.3) | |
10 mm |
50 mm | |
5/Pk. |