Learn More
WTW™ Glass Overflow Measuring Flask
Overflow measuring flasks are a simple and very precise way to conform to fixed volume instructions.
Brand: WTW™ 209050
495.93 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
97 mL | |
BOD | |
Glass |
Product Content Correction
Product Title
WTW™ Glass Overflow Measuring Flask > Capacity: 97mL
Spot an opportunity for improvement?Share a Content Correction