Learn More
Macherey-Nagel™ Glass TLC Plate
Product Code. 15278078 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
25 Pk.
Unit Size:
Each
Product Code. Quantity Unit Size
15278078 25 Pk. Each
1 options
Product Code. 15278078 Supplier Macherey-Nagel™ Supplier No. 810123
  • ONLINE EXCLUSIVE PRICE
    3160.00 SEK
    2580.13 SEK / Paket med 1
    Save 579.87 SEK
    18% Av
View all promotions
Beräknad leverans: 07-12-2026
TLC glass plates in different plate sizes for many different applications, SILGUR-25-C (channel plate), Unmodified silica gel (SiOH) with concentrating zone (kieselguhr, diatomaceous earth or diatomite).
Specifications
Material Glass
Quantity 25 Pk.
Type TLC Plate
Fluorescent Indicator UV254
For Use With (Equipment) 19 Channels
Includes Channels
Thickness (Metric) 0.25 mm