missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLDN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26185-1-AP-20UL
This item is not returnable.
View return policy
Description
CRG-L2 is a protein that contains olfactomedin-like and collagen-like domains. The encoded protein, which exists in both transmembrane and secreted forms, promotes formation of the nodes of Ranvier in the peripheral nervous system. Mutations in this gene cause a form of lethal congenital contracture syndrome in human patients. Autoantibodies to the encoded protein have been identified in sera form patients with multifocal motor neuropathy.Specifications
GLDN | |
Polyclonal | |
Unconjugated | |
GLDN | |
CLOM, COLM, CRG L2, CRGL2, GLDN, gliomedin, UNC 112 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235379, 342035 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZMI3, Q8BMF8 | |
GLDN | |
GLDN Fusion Protein Ag24247 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLDN Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction