missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GLI4 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896204
|
Proteintech
16551-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886204
|
Proteintech
16551-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLI4 is a protein coding gene.Specifications
GLI4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P10075 | |
100043133, 2738, 500893 | |
GLI4 Fusion Protein Ag9497 | |
Primary | |
-20°C | |
GLI4 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GLI family zinc finger 4, GLI4, HKR4, Protein HKR4, Zinc finger protein GLI4 | |
9130023H24Rik, GLI4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech