missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLIS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23138-1-AP-20UL
This item is not returnable.
View return policy
Description
MAP2K1 (MEK1) is a dual specificity mitogen-activated protein kinase (MAPK) kinase. It is an essential component of the MAP kinase pathway and stimulates the enzymatic activity of MAP kinases in response to various signals. Growth factors, cytokines, and proto-oncogenes transduce their signals through the activation of RAS, which leads to activation of the serine/threonine kinase RAF, and then to activation of MAPK kinase. MAP2K1 is activated by the Raf family member p74raf-1, through phosphorylation of serine residues at positions 217 and 221 in the activation loop.Specifications
GLIS1 | |
Polyclonal | |
Unconjugated | |
GLIS1 | |
GLI similar 1, GLIS family zinc finger 1, GLIS1, Zinc finger protein GLIS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
148979 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NBF1 | |
GLIS1 | |
GLIS1 Fusion Protein Ag19214 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLIS1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction