missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP1R Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26196-1-AP-20UL
This item is not returnable.
View return policy
Description
Glucagon-like peptide-1(9-36)amide and GLP-1(9-37) are the forms of GLP-1 that result from the rapid degradation of the active forms of the peptide (GLP-1(7-36)amide and GLP-1(7-37)) by the enzyme dipeptidyl peptidase-IV (DPP-IV, also known as CD26 or adenosine deaminase binding protein). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion. The sequence of GLP-1 is fully conserved in all mammalian species examined so far.Specifications
GLP1R | |
Polyclonal | |
Unconjugated | |
GLP1R | |
GLP 1 R, GLP 1 receptor, GLP 1R, GLP1R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14652, 25051, 2740 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35659, P32301, P43220 | |
GLP1R | |
GLP1R Fusion Protein Ag23529 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLP1R Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction