missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14299-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a subunit of glutamate receptor channels. These channels mediate most of the fast excitatory synaptic transmission in the central nervous system and play key roles in synaptic plasticity.Specifications
GLUD1 | |
Polyclonal | |
Unconjugated | |
GLUD1 | |
GDH, GDH 1, GDH1, GLUD, GLUD1, glutamate dehydrogenase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14661, 24399, 2746 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00367, P10860, P26443 | |
GLUD1 | |
GLUD1 Fusion Protein Ag5694 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLUD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction