missing translation for 'onlineSavingsMsg'
Learn More

GLUT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen GLUT3
Concentration 0.33 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16879934
View Documents
Proteintech
20403-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16869934
View Documents
Proteintech
20403-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody was generated against the internal region of human GLUT3 and may cross-react with GLUT14.20403-1-AP antibody detects the bands around 48-60 kDa in SDS-PAGE.

Glucose is the major source of our energy and there are numerous isoforms of the glucose transporter in mammals, including Glut1, Glut2, Glut3, Glut4, Glut5, Glut6, Glut7, Glut8 and Glut9. The Glut5 gene located on the short arm of human chromosome 1 encodes a 501-amino acid facilitative glucose transporter. Glut5 mRNA is highly expressed in small intestine and to a lesser extent in kidney, skeletal muscle and adipose tissue. Glut5 plays a critical role in fructose absorption in the small intestine and its expression is highly induced when exposed to a fructose-enriched diet. Glut5 transporter expressed in human skeletal muscle is specifically localized to the plasma membrane, where it participates in regulating hexose transfer across the sarcolemma. Glut8, a novel glucose transporter-like protein, exhibits significant sequence similarity with the other members of sugar transporter family. Glut8 comprises 12 putative membrane-spanning helices and several conserved motifs, which are important for transport activity. In human tissues, Glut8 is predomitly expressed in testis and, to a lesser extent, in most other tissues including skeletal muscle, heart, small intestine and brain.
Specifications

Specifications

GLUT3
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse
P11169, P32037
20527, 6515
GLUT3 Fusion Protein Ag14203
Primary
-20°C
SLC2A3
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GLUT 3, GLUT3, SLC2A3
SLC2A3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GLUT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.