Learn More
GLUT5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27571-1-AP-20UL
 View more versions of this product
Product Code. 16885705
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
SLC2A5 (GLUT5) is a cytochalasin B-sensitive carrier. It seems to function primarily as a fructose transporter.
Specifications
Specifications
GLUT5
Polyclonal
Unconjugated
SLC2A5
Fructose transporter, GLUT 5, GLUT5, SLC2A5
Rabbit
Antigen Affinity Chromatography
RUO
56485, 6518
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22732, Q9WV38
SLC2A5
GLUT5 Fusion Protein Ag25682
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GLUT5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated