Learn More
GLUT5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27571-1-AP-20UL
 View more versions of this product
Product Code. 16885705
  • 1645.54 SEK / 20µL
Description
SLC2A5 (GLUT5) is a cytochalasin B-sensitive carrier. It seems to function primarily as a fructose transporter.
Specifications
Specifications
GLUT5
Polyclonal
Unconjugated
SLC2A5
Fructose transporter, GLUT 5, GLUT5, SLC2A5
Rabbit
Antigen Affinity Chromatography
RUO
56485, 6518
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22732, Q9WV38
SLC2A5
GLUT5 Fusion Protein Ag25682
20 μL
Primary
Mouse, Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GLUT5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated