missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamate receptor 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Glutamate receptor 2 |
---|---|
Concentration | 0.42 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879433
|
Proteintech
11994-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869433
|
Proteintech
11994-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain (Keien et al., 1990; Hollmann and Heinemann, 1994). The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD (Chung et al., 2003).Specifications
Glutamate receptor 2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P19491, P23819, P42262 | |
14800, 2891, 29627 | |
Glutamate receptor 2 Fusion Protein Ag2662 | |
Primary | |
-20°C | |
GRIA2 |
0.42 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GluA2, GluR 2, GluR B, GluR K2, GLUR2, GLURB, Glutamate receptor 2, GRIA2, HBGR2 | |
GRIA2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glutamate receptor 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech