missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GMDS |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894614
|
Proteintech
15442-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884614
|
Proteintech
15442-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyzes the conversion of GDP-D-mannose to GDP-4-dehydro-6-deoxy-D-mannose.Specifications
GMDS | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O60547, Q8K0C9 | |
218138, 2762, 291095 | |
GMDS Fusion Protein Ag7319 | |
Primary | |
-20°C | |
GMDS |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GDP D mannose dehydratase, GDP mannose 4,6 dehydratase, GMD, GMDS, SDR3E1 | |
GMDS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GMDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech