Learn More
GNA13 Mouse anti-Human, Unconjugated, Clone: 2G8F10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67188-1-IG-20UL
 View more versions of this product
Product Code. 16889685
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The GNA13 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNA13. This antibody recognizes human, mouse, rat, pig antigen. The GNA13 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GNA13
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry
2G8F10
Unconjugated
20 μL
10672
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GNA13 Mouse anti-Human, Unconjugated, Clone: 2G8F10, Proteintech > 20 μL