Learn More
GNA13 Mouse anti-Human, Unconjugated, Clone: 2G8F10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67188-1-IG-20UL
 View more versions of this product
Product Code. 16889685
  • 1645.54 SEK / 20µL
Description
The GNA13 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNA13. This antibody recognizes human, mouse, rat, pig antigen. The GNA13 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GNA13
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry
2G8F10
Unconjugated
20 μL
10672
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GNA13 Mouse anti-Human, Unconjugated, Clone: 2G8F10, Proteintech > 20 μL