Learn More
GNA13 Mouse anti-Human, Unconjugated, Clone: 2G8F10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67188-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879685
  • 3665.00 SEK / 150µL
Estimated Shipment: 28-03-2023
to see stock.
Description
Description
The GNA13 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNA13. This antibody recognizes human, mouse, rat, pig antigen. The GNA13 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GNA13
Monoclonal
1000 μg/mL
Mouse
Primary
Pig, Rat, Mouse, Human
Immunohistochemistry, Western Blot
2G8F10
Unconjugated
150 μL
10672
IgG1