Learn More
GNB3 Mouse anti-Human, Unconjugated, Clone: 2B6E1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GNB3 |
---|---|
Clone | 2B6E1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880156
|
Proteintech
67497-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870156
|
Proteintech
67497-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GNB3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNB3. This antibody recognizes human, mouse, rat, pig antigen. The GNB3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
GNB3 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
2784 | |
Primary |
2B6E1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.