Learn More
GNB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10081-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Heterotrimeric guanine nucleotide-binding proteins in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.Specifications
GNB3 | |
Polyclonal | |
Unconjugated | |
GNB3 | |
GNB3, Transducin beta chain 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14695, 2784, 60449 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16520, Q61011 | |
GNB3 | |
GNB3 Fusion Protein Ag0127 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |