Learn More
GNRHR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20728-1-AP-20UL
 View more versions of this product
Product Code. 16890295
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
In non-hominoid primates and non-mammalian vertebrates, the gonadotropin releasing hormone 2 receptor gene (GnRHR2) encodes a seven-transmembrane G-protein coupled receptor. However, in human, the corresponding reading frame contains a premature stop codon, which has been suggested to encode a selenocysteine residue, but there is no solid evidence for selenocysteine incorporation. It appears that the human GnRHR2 transcription occurs but the gene does not likely produce a functional multi-transmembrane protein. A non-transcribed pseudogene of GnRHR2 is located on chromosome 14. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
GNRHR2
Polyclonal
Unconjugated
GNRHR2
GnRH II R, GNRHR2
Rabbit
Antigen Affinity Chromatography
RUO
114814
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96P88
GNRHR2
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GNRHR2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated