missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RER1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RER1.
Brand: Abnova PAB7598.100ug
This item is not returnable.
View return policy
Description
Sequence: C-HGKRRYRGKEDAGKSpecifications
RER1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) The optimal working dilution should be determined by the end user. | |
RER1 | |
Goat | |
Antigen affinity purification | |
RUO | |
11079 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of RER1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
RER1 | |
A synthetic peptide corresponding to human RER1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RER1, Goat, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction