missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21193-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation.Specifications
GOLGA3 | |
Polyclonal | |
Unconjugated | |
GOLGA3 | |
GCP170, GOLGA3, Golgin 160, Golgin subfamily A member 3, MEA 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269682, 2802 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55937, Q08378 | |
Golga3 | |
GOLGA3 Fusion Protein Ag15531 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GOLGA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction