missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOSR2/Membrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12095-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
GOSR2/Membrin | |
Polyclonal | |
Unconjugated | |
GOSR2 | |
27 kDa Golgi SNARE protein, Bos1, GOSR2, GS27, Membrin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56494, 9570 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14653, O35166 | |
Gosr2 | |
GOSR2/Membrin Fusion Protein Ag2736 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GOSR2/Membrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction