Learn More
A&D Weighing GP Series Precision Balances Internal Calibration
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
52760.00 SEK - 70970.00 SEK
Specifications
Display Type | VFD |
---|---|
Stabilization Time | 1.5 seconds |
Type | Industrial Precision Balance |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16641132
![]() |
A&D Weighing
GP-40K |
41 kg |
52760.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16671132
![]() |
A&D Weighing
GP-61K |
61 kg |
70970.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Swing arm design or separate display and platform design
- IP65 protected
- Durable aluminum alloy & stainless steel construction
- Easy calibration with internal calibration mass
- SHS response speed of only 1.5 seconds
- GLP, GMP and ISO compliant
- RS-232C interface
- Comparator output and analog output (optional)
- Large Vacuum Fluorescent display
- Data memory function
- Counting function with ACAI (Automatic Counting Accuracy Improvement)
- Comparator function
- Underhook weighing for thermal and magnetic samples as well as for density calculation
- Animal weighing function
Specifications
VFD | |
1.5 seconds | |
Industrial Precision Balance |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
A&D Weighing GP Series Precision Balances Internal Calibration