Learn More
GP73/GOLPH2 Mouse anti-Human, Clone: 3D1F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66331-1-IG-150UL
 View more versions of this product
Product Code. 16888105
  • 4652.90 SEK / 150µL
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes protein synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this encoded protein has been observed to be upregulated in response to viral infection.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GP73/GOLPH2 Mouse anti-Human, Clone: 3D1F1, Proteintech > 150 μL; Unconjugated