Learn More
GPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13451-1-AP-20UL
 View more versions of this product
Product Code. 16881674
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the NAD-dependent glycerol-3-phosphate dehydrogenase family. The encoded protein plays a critical role in carbohydrate and lipid metabolism by catalyzing the reversible conversion of dihydroxyacetone phosphate (DHAP) and reduced nicotine adenine dinucleotide (NADH) to glycerol-3-phosphate (G3P) and NAD+. The encoded cytosolic protein and mitochondrial glycerol-3-phosphate dehydrogenase also form a glycerol phosphate shuttle that facilitates the transfer of reducing equivalents from the cytosol to mitochondria. Mutations in this gene are a cause of transient infantile hypertriglyceridemia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
GPD1
Polyclonal
Unconjugated
GPD1
cGPDH, GPD C, GPD1, GPDH C
Rabbit
Antigen Affinity Chromatography
RUO
14555, 2819, 60666
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35077, P13707, P21695
GPD1
GPD1 Fusion Protein Ag4278
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated