Learn More
GPD1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17263-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene catalyzes the conversion of sn-glycerol 3-phosphate to glycerone phosphate. The encoded protein is found in the cytoplasm, associated with the plasma membrane, where it binds the sodium channel, voltage-gated, type V, alpha subunit (SCN5A). Defects in this gene are a cause of Brugada syndrome type 2 (BRS2) as well as sudden infant death syndrome (SIDS).Specifications
GPD1L | |
Polyclonal | |
Unconjugated | |
GPD1L | |
GPD1 L, GPD1L, KIAA0089 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23171, 333433, 363159 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3ULJ0, Q8N335 | |
Gpd1l | |
GPD1L Fusion Protein Ag10977 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
GPD1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction