Learn More
GPNMB Mouse anti-Human, Unconjugated, Clone: 2B10B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66926-1-IG-150UL
 View more versions of this product
Product Code. 16879225
  • 4652.90 SEK / 150µL
Description
The GPNMB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GPNMB. This antibody recognizes human, mouse, rat antigen. The GPNMB antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
GPNMB
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2B10B8
Unconjugated
150 μL
10457
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GPNMB Mouse anti-Human, Unconjugated, Clone: 2B10B8, Proteintech > 150 μL