missing translation for 'onlineSavingsMsg'
Learn More

GPR126 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen GPR126
Applications Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897744
View Documents
Proteintech
17774-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16887744
View Documents
Proteintech
17774-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The GPR126 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR126. This antibody recognizes human, mouse, rat antigen. The GPR126 antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Specifications

GPR126
Polyclonal
Rabbit
57211
Primary
Immunohistochemistry, ELISA
Unconjugated
Human, Rat, Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
GPR126 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.