missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR183/EBI2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12377-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
GPR183 was identified by the up-regulation of its expression upon Epstein-Barr virus infection of primary B lymphocytes. GPR183 is predicted to encode a G protein-coupled receptor that is most closely related to the thrombin receptor. Expression of this gene was detected in B-lymphocyte cell lines and lymphoid tissues but not in T-lymphocyte cell lines or peripheral blood T lymphocytes. The function of this gene is unknown.Specifications
GPR183/EBI2 | |
Polyclonal | |
Unconjugated | |
GPR183 | |
EBI2, G protein coupled receptor 183, GP183, GPR183 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1880, 321019 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P32249, Q3U6B2 | |
Gpr183 | |
GPR183/EBI2 Fusion Protein Ag3035 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |